The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271397.1 | NP_001258326.1 | 13 - 14 | 3739 | 104 |
NR_073167.1 | 13 - 14 | 3920 | 104 | ||
NR_073168.1 | 13 - 14 | 3735 | 104 | ||
GenBank mRNA | AK147172.1 | - | 13 - 14 | 3475 | 104 |
BC026402.1 | - | 2 - 3 | 356 | 104 | |
BC058699.1 | - | 13 - 14 | 3895 | 104 | |
BC094941.1 | - | 5 - 6 | 609 | 104 | |
BC125575.1 | - | 13 - 14 | 3465 | 104 | |
BC125579.1 | - | 13 - 14 | 3465 | 104 |