Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Riok1
Entrez Gene ID
71340
Gene Name
RIO kinase 1 (yeast)
Gene Aliases
3110046C13Rik, 5430416A05Rik, Ad034
Chromosome Location
Chr.13: 38036989 - 38061433 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024242.3 NP_077204.2 15 - 16 1913 129
GenBank mRNA AK017312.1 - 15 - 16 1608 129
AK152707.1 - 15 - 16 1662 129
AK161315.1 - 16 - 17 2156 129
BC002158.1 - 9 - 10 870 129

View Full Product Details