Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zcwpw2
Entrez Gene ID
100039681
Gene Name
zinc finger, CW type with PWWP domain 2
Gene Aliases
4930430K04Rik
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK015254.1 - - 289 75

View Full Product Details