Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Larp1
- Entrez Gene ID
-
73158
- Gene Name
- La ribonucleoprotein domain family, member 1
- Gene Aliases
-
1810024J12Rik, 3110040D16Rik, Larp, mKIAA0731
- Chromosome Location
-
Chr.11: 58009064 - 58062034 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_028451.1 |
NP_082727.1 |
2 - 3 |
545 |
66 |
View Full Product Details