The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009354.1 | NP_033380.1 | 13 - 14 | 3038 | 88 |
GenBank mRNA | AF051911.1 | - | 13 - 14 | 3038 | 88 |
AF073311.1 | - | 13 - 14 | 3009 | 88 | |
BC082327.1 | - | 14 - 15 | 2540 | 88 | |
BC127068.1 | - | 14 - 15 | 2326 | 88 |