The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271873.1 | NP_001258802.1 | 12 - 13 | 1873 | 72 |
NM_133979.3 | NP_598740.1 | 12 - 13 | 2047 | 72 | |
GenBank mRNA | AK028391.1 | - | 7 - 8 | 991 | 72 |
AK046091.1 | - | 12 - 13 | 1999 | 72 | |
AK080947.1 | - | 12 - 13 | 1999 | 72 | |
AK219476.1 | - | 1 - 2 | 115 | 72 | |
BC002294.1 | - | 3 - 4 | 449 | 72 | |
BC026421.1 | - | 7 - 8 | 1087 | 72 | |
BC099688.1 | - | 12 - 13 | 1796 | 72 |