The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271873.1 | NP_001258802.1 | 1 - 2 | 117 | 95 |
NM_133979.3 | NP_598740.1 | 1 - 2 | 117 | 95 | |
GenBank mRNA | AK043011.1 | - | 1 - 2 | 80 | 95 |
AK046091.1 | - | 1 - 2 | 69 | 95 | |
AK079622.1 | - | 1 - 2 | 68 | 95 | |
AK080947.1 | - | 1 - 2 | 69 | 95 | |
AK138414.1 | - | 1 - 2 | 103 | 95 |