Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ell
Entrez Gene ID
13716
Gene Name
elongation factor RNA polymerase II
Gene Aliases
Ell1, Men
Chromosome Location
Chr.8: 70539675 - 70592858 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007924.2 NP_031950.2 8 - 9 1444 72
GenBank mRNA AK159027.1 - 8 - 9 1431 72
BC014816.1 - 8 - 9 1414 72
BC024894.1 - 8 - 9 1413 72
U80227.1 - 8 - 9 1417 72

View Full Product Details