Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sdha
Entrez Gene ID
66945
Gene Name
succinate dehydrogenase complex, subunit A, flavoprotein (Fp)
Gene Aliases
1500032O14Rik, 2310034D06Rik, 4921513A11, C81073, FP, SDH2, SDHF
Chromosome Location
Chr.13: 74322255 - 74350240 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023281.1 NP_075770.1 2 - 3 174 67
GenBank mRNA AK010561.1 - 2 - 3 163 67
AK029520.1 - 2 - 3 174 67
AK034928.1 - 2 - 3 175 67
AK049590.1 - 2 - 3 161 67
AK050475.1 - 2 - 3 161 67
AK075990.1 - 2 - 3 158 67
AK145923.1 - 2 - 3 172 67
AK147286.1 - 2 - 3 161 67
AK147624.1 - 2 - 3 161 67
AK153085.1 - 2 - 3 181 67
AK162148.1 - 2 - 3 157 67
AK169254.1 - 2 - 3 157 67
AK207923.1 - 1 - 2 50 67
BC011301.1 - 2 - 3 134 67
BC031849.1 - 2 - 3 182 67

View Full Product Details