The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028959.3 | NP_083235.3 | 11 - 12 | 1804 | 64 |
GenBank mRNA | AK011607.1 | - | 6 - 7 | 728 | 64 |
AK017134.1 | - | 11 - 12 | 1796 | 64 | |
AK034892.1 | - | 11 - 12 | 1797 | 64 | |
AK161033.1 | - | 7 - 8 | 1129 | 64 | |
AK173189.1 | - | 4 - 5 | 3788 | 64 | |
BC100549.1 | - | 10 - 11 | 1654 | 64 |