Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cep72
Entrez Gene ID
74470
Gene Name
centrosomal protein 72
Gene Aliases
2610029E11Rik, 4933440J22Rik
Chromosome Location
Chr.13: 74036495 - 74062314 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028959.3 NP_083235.3 4 - 5 532 95
GenBank mRNA AK017134.1 - 4 - 5 524 95
AK034892.1 - 4 - 5 525 95
BC060686.1 - 4 - 5 530 95
BC080681.1 - 4 - 5 488 95
BC100549.1 - 3 - 4 382 95

View Full Product Details