Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc9a3
Entrez Gene ID
105243
Gene Name
solute carrier family 9 (sodium/hydrogen exchanger), member 3
Gene Aliases
9030624O13Rik, AI930210, NHE-3, NHE3
Chromosome Location
Chr.13: 74086254 - 74169442 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081060.1 NP_001074529.1 10 - 11 1636 106

View Full Product Details