Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Otud5
Entrez Gene ID
54644
Gene Name
OTU domain containing 5
Gene Aliases
AA407879, AI553596, BB114028, DUBA, DXImx46e, Sfc7
Chromosome Location
Chr.X: 7841831 - 7876626 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290537.1 NP_001277466.1 1 - 2 393 99
GenBank mRNA AK010750.1 - 1 - 2 391 99
AK149777.1 - 1 - 2 394 99
AK169921.1 - 1 - 2 359 99

View Full Product Details