The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290430.1 | NP_001277359.1 | 12 - 13 | 1256 | 95 |
NM_133991.2 | NP_598752.2 | 12 - 13 | 1262 | 95 | |
GenBank mRNA | AK036313.1 | - | 12 - 13 | 1220 | 95 |
BC011144.1 | - | 11 - 12 | 1118 | 95 | |
BC034880.1 | - | 12 - 13 | 1126 | 95 |