Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Spata9
Entrez Gene ID
75571
Gene Name
spermatogenesis associated 9
Gene Aliases
1700030K01Rik, 4930599C08Rik, A930023H06Rik
Chromosome Location
Chr.13: 75960003 - 75998987 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029343.3 NP_083619.1 3 - 4 471 153
GenBank mRNA AK006554.1 - 3 - 4 472 153
BC048558.1 - 3 - 4 451 153

View Full Product Details