The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138953.2 | NP_620403.2 | 3 - 4 | 487 | 100 |
GenBank mRNA | AK140906.1 | - | 3 - 4 | 441 | 100 |
AK145868.1 | - | 3 - 4 | 436 | 100 | |
AK154545.1 | - | 3 - 4 | 448 | 100 | |
AK156164.1 | - | 3 - 4 | 453 | 100 | |
BC151188.1 | - | 3 - 4 | 442 | 100 |