The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001131068.1 | NP_001124540.1 | 2 - 3 | 763 | 157 |
NM_001131069.1 | NP_001124541.1 | 2 - 3 | 226 | 157 | |
NM_178916.5 | NP_849247.3 | 2 - 3 | 1213 | 157 | |
GenBank mRNA | AK029602.1 | - | 2 - 3 | 1038 | 157 |
AK040255.1 | - | 2 - 3 | 718 | 157 | |
AK083705.1 | - | 2 - 3 | 1214 | 157 | |
BC030381.1 | - | 2 - 3 | 838 | 157 | |
CD769672.1 | - | 2 - 3 | 230 | 157 | |
CN839985.1 | - | 2 - 3 | 236 | 157 |