The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040459.1 | NP_001035549.1 | 8 - 9 | 4160 | 62 |
GenBank mRNA | AK173129.1 | - | 7 - 8 | 4182 | 62 |
BC151022.1 | - | 8 - 9 | 4451 | 62 | |
DQ435686.1 | - | 8 - 9 | 4160 | 62 | |
DQ435687.1 | - | 8 - 9 | 4160 | 62 |