The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166397.1 | NP_001159869.1 | 1 - 2 | 304 | 131 |
NM_026139.4 | NP_080415.3 | 1 - 2 | 304 | 131 | |
GenBank mRNA | AK034185.1 | - | 1 - 2 | 80 | 131 |
AK034621.1 | - | 1 - 2 | 78 | 131 | |
AK043440.1 | - | 1 - 2 | 80 | 131 | |
AK044926.1 | - | 1 - 2 | 78 | 131 | |
AK053354.1 | - | 1 - 2 | 78 | 131 | |
AK172960.1 | - | 1 - 2 | 304 | 131 | |
BC049088.1 | - | 1 - 2 | 59 | 131 | |
BC052383.1 | - | 1 - 2 | 53 | 131 |