Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cyp4f41-ps
Entrez Gene ID
77875
Gene Name
cytochrome P450, family 4, subfamily f, polypeptide 41 pseudogene
Gene Aliases
6030422M01Rik
Chromosome Location
Chr.17: 32950961 - 32965716 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_033585.1 1 - 2 52 64
GenBank mRNA AK020053.1 - 1 - 2 36 64

View Full Product Details