The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023065.3 | NP_075552.2 | 2 - 3 | 337 | 84 |
GenBank mRNA | AF290973.1 | - | 2 - 3 | 338 | 84 |
AF309649.1 | - | 2 - 3 | 305 | 84 | |
AK077725.1 | - | 2 - 3 | 338 | 84 | |
AK156914.1 | - | 2 - 3 | 338 | 84 | |
BC054852.1 | - | 2 - 3 | 323 | 84 | |
BQ746455.1 | - | 2 - 3 | 293 | 85 |