Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hrct1
Entrez Gene ID
100039781
Gene Name
histidine rich carboxyl terminus 1
Gene Aliases
3830408D24Rik
Chromosome Location
Chr.4: 43727198 - 43728110 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK014425.1 - - 1140 126

View Full Product Details