Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cmtm6
Entrez Gene ID
67213
Gene Name
CKLF-like MARVEL transmembrane domain containing 6
Gene Aliases
2810051A14Rik, AA536733, Cklfsf6
Chromosome Location
Chr.9: 114731157 - 114749347 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026036.3 NP_080312.1 1 - 2 177 70
GenBank mRNA AK012941.1 - 1 - 2 178 70
AK030885.1 - 1 - 2 157 70
AK128960.1 - 1 - 2 163 70
AK150775.1 - 1 - 2 178 70
AK151092.1 - 1 - 2 178 70
AK151431.1 - 1 - 2 178 70
AK152448.1 - 1 - 2 179 70
AK152723.1 - 1 - 2 188 70
AK152817.1 - 1 - 2 188 70
AK153092.1 - 1 - 2 178 70
AK154481.1 - 1 - 2 164 70
AK159439.1 - 1 - 2 147 70
AK159735.1 - 1 - 2 172 70
AK165948.1 - 1 - 2 146 70
AK167499.1 - 1 - 2 164 70
AK167854.1 - 1 - 2 169 70
AK170905.1 - 1 - 2 179 70
AK171394.1 - 1 - 2 208 70
AY241868.1 - 1 - 2 214 70
BC027248.1 - 1 - 2 149 70

View Full Product Details