Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Secisbp2
Entrez Gene ID
75420
Gene Name
SECIS binding protein 2
Gene Aliases
2210413N07Rik, 2810012K13Rik, SBP2
Chromosome Location
Chr.13: 51650829 - 51684044 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK012732.1 - 7 - 8 1137 109
AK030021.1 - 7 - 8 1107 109
AK156076.1 - 7 - 8 1183 109
AK168634.1 - 7 - 8 1140 109
BC125391.1 - 7 - 8 1163 109
BC125393.1 - 7 - 8 1160 109
BC144795.1 - 7 - 8 1158 109

View Full Product Details