Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fbxo42
Entrez Gene ID
213499
Gene Name
F-box protein 42
Gene Aliases
6720460I06Rik, mKIAA1332
Chromosome Location
Chr.4: 141147907 - 141204062 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172518.3 NP_766106.2 2 - 3 520 83
GenBank mRNA AK028867.1 - 2 - 3 366 83
AK150804.1 - 2 - 3 511 83
AK151503.1 - 2 - 3 477 83
AK165284.1 - 2 - 3 494 83
BC058667.1 - 2 - 3 511 83

View Full Product Details