Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tspan6
Entrez Gene ID
56496
Gene Name
tetraspanin 6
Gene Aliases
6720473L21Rik, AI316786, T245, TSPAN-6, Tm4sf, Tm4sf6
Chromosome Location
Chr.X: 133891070 - 133898429 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019656.3 NP_062630.2 3 - 4 476 86
GenBank mRNA AF053454.1 - 3 - 4 423 86
AK146673.1 - 3 - 4 381 86
AK149204.1 - 3 - 4 386 86
AK160313.1 - 3 - 4 370 86
AK167384.1 - 3 - 4 372 86
BC003733.1 - 3 - 4 410 86
BC110314.1 - 3 - 4 385 86

View Full Product Details