The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009427.2 | NP_033453.2 | 2 - 2 | 1319 | 77 |
GenBank mRNA | AK142667.1 | - | 2 - 2 | 1338 | 77 |
BC082588.1 | - | 2 - 2 | 1319 | 77 |