Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Atxn7l1
Entrez Gene ID
380753
Gene Name
ataxin 7-like 1
Gene Aliases
2810423G08Rik, Atxn7l4
Chromosome Location
Chr.12: 33147639 - 33376408 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC100598.1 - 2 - 3 217 58
BY203850.1 - 3 - 4 277 58

View Full Product Details