Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Trps1
- Entrez Gene ID
-
83925
- Gene Name
- trichorhinophalangeal syndrome I (human)
- Gene Aliases
-
AI115454, AI447310, D15Ertd586e
- Chromosome Location
-
Chr.15: 50654757 - 50890747 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC037058.1 |
- |
6 - 7 |
2213 |
69 |
View Full Product Details