The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029609.1 | NP_083885.1 | 6 - 7 | 782 | 82 |
GenBank mRNA | AK009207.1 | - | 6 - 7 | 780 | 82 |
AK140229.1 | - | 1 - 2 | 71 | 82 |