The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033466.3 | NP_001028638.2 | 2 - 3 | 449 | 93 |
GenBank mRNA | AK030936.1 | - | 2 - 3 | 297 | 93 |
AK132308.1 | - | 2 - 3 | 450 | 93 | |
AK160748.1 | - | 2 - 3 | 279 | 93 |