Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cyp3a59
Entrez Gene ID
100041449
Gene Name
cytochrome P450, family 3, subfamily a, polypeptide 59
Gene Aliases
ENSMUSG00000061292
Chromosome Location
Chr.5: 146079209 - 146113791 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105160.1 NP_001098630.1 4 - 5 396 123

View Full Product Details