The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009460.2 | NP_033486.1 | 1 - 2 | 153 | 145 |
GenBank mRNA | AF033353.1 | - | 1 - 2 | 128 | 145 |
AK002536.1 | - | 1 - 2 | 123 | 145 | |
AK011074.1 | - | 1 - 2 | 122 | 144 | |
AK159366.1 | - | 1 - 2 | 124 | 145 | |
BC082566.1 | - | 1 - 2 | 128 | 145 | |
BC083158.1 | - | 1 - 2 | 126 | 145 | |
BY242724.1 | - | 1 - 2 | 154 | 145 |