The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_104434.1 | 1 - 2 | 271 | 143 | |
GenBank mRNA | AK010140.1 | - | 1 - 2 | 223 | 143 |
AK010864.1 | - | 1 - 2 | 266 | 143 | |
AK168268.1 | - | 1 - 2 | 252 | 143 |