Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Try10
Entrez Gene ID
436522
Gene Name
trypsin 10
Gene Aliases
-
Chromosome Location
Chr.6: 41354105 - 41357944 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038996.2 NP_001034085.1 2 - 3 290 151
GenBank mRNA AK133516.1 - 2 - 3 217 151
BC138698.1 - 2 - 3 290 151
BC146017.1 - 2 - 3 290 151

View Full Product Details