The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166157.1 | NP_001159629.1 | 7 - 8 | 1316 | 148 |
GenBank mRNA | AF312018.1 | - | 7 - 8 | 977 | 148 |