The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010798.2 | NP_034928.1 | 2 - 3 | 363 | 177 |
GenBank mRNA | AK008939.1 | - | 2 - 3 | 357 | 177 |
AK010691.1 | - | 2 - 3 | 334 | 177 | |
AK013328.1 | - | 2 - 3 | 357 | 177 | |
AK152211.1 | - | 2 - 3 | 177 | ||
BC024895.1 | - | 2 - 3 | 343 | 177 | |
BC086928.1 | - | 2 - 3 | 333 | 177 | |
BC100605.1 | - | 2 - 3 | 333 | 177 | |
L02913.1 | - | 2 - 3 | 336 | 177 | |
L07607.1 | - | 2 - 3 | 266 | 177 | |
L10613.1 | - | 2 - 3 | 362 | 177 | |
Z23048.1 | - | 2 - 3 | 283 | 177 |