Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rpl13
Entrez Gene ID
270106
Gene Name
ribosomal protein L13
Gene Aliases
A52, L13
Chromosome Location
Chr.8: 123102350 - 123105244 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016738.5 NP_058018.2 1 - 2 101 104
GenBank mRNA CF751804.1 - 1 - 2 154 104

View Full Product Details