The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013570.1 | NP_038598.1 | 1 - 2 | 385 | 131 |
GenBank mRNA | AA681979.1 | - | 1 - 2 | 385 | 131 |
AI466315.1 | - | 1 - 2 | 126 | 131 |