Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Serpina3i
Entrez Gene ID
628900
Gene Name
serine (or cysteine) peptidase inhibitor, clade A, member 3I
Gene Aliases
2B2, EG628900, Gm6930
Chromosome Location
Chr.12: 104263045 - 104269365 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199940.1 NP_001186869.1 4 - 5 1118 112

View Full Product Details