The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013725.4 | NP_038753.1 | 3 - 4 | 299 | 78 |
GenBank mRNA | AK005147.1 | - | 3 - 4 | 244 | 78 |
AK150738.1 | - | 3 - 4 | 274 | 78 | |
BC012641.1 | - | 3 - 4 | 313 | 78 | |
CJ044201.1 | - | 3 - 4 | 300 | 78 | |
U93864.1 | - | 3 - 4 | 229 | 78 |