The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198027.2 | NP_932144.2 | 6 - 7 | 561 | 61 |
GenBank mRNA | AK082058.1 | - | 5 - 6 | 729 | 61 |
AK209174.1 | - | 2 - 3 | 209 | 61 | |
BC029805.1 | - | 6 - 7 | 513 | 61 | |
BC080310.1 | - | 4 - 5 | 684 | 61 |