Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gp49a
Entrez Gene ID
14727
Gene Name
glycoprotein 49 A
Gene Aliases
gp49
Chromosome Location
Chr.10: 51480612 - 51486329 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291892.1 NP_001278821.1 5 - 6 811 165
NM_001291893.1 NP_001278822.1 4 - 5 521 165
NM_008147.2 NP_032173.1 5 - 6 806 165
GenBank mRNA AK089366.1 - 4 - 5 358 165
AK167751.1 - 5 - 6 727 165
AK171573.1 - 5 - 6 755 166
BC065784.1 - 4 - 5 475 165
M65027.1 - 5 - 6 739 165

View Full Product Details