The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010373.3 | NP_034503.2 | 2 - 3 | 272 | 163 |
GenBank mRNA | BG146331.1 | - | 2 - 3 | 312 | 163 |
J03256.1 | - | 2 - 3 | 184 | 163 | |
M36901.1 | - | 2 - 3 | 164 | 163 | |
X14093.1 | - | 2 - 3 | 164 | 163 |