Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Snrpg
- Entrez Gene ID
-
68011
- Gene Name
- small nuclear ribonucleoprotein polypeptide G
- Gene Aliases
-
2810024K17Rik, AL022803, SMG, sm-G, snRNP-G
- Chromosome Location
-
Chr.6: 86371540 - 86378902 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC051470.1 |
- |
1 - 2 |
113 |
130 |
View Full Product Details