Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vmn2r8
Entrez Gene ID
627479
Gene Name
vomeronasal 2, receptor 8
Gene Aliases
EG627479
Chromosome Location
Chr.5: 108797193 - 108808754 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104620.1 NP_001098090.1 2 - 3 480 125

View Full Product Details