Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vmn2r74
Entrez Gene ID
546980
Gene Name
vomeronasal 2, receptor 74
Gene Aliases
EG546980
Chromosome Location
Chr.7: 85951867 - 85961482 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105187.1 NP_001098657.1 5 - 6 1650 139

View Full Product Details