Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Serf2
Entrez Gene ID
378702
Gene Name
small EDRK-rich factor 2
Gene Aliases
C80083, Msmac1l, m4F5rel
Chromosome Location
Chr.2: 121449198 - 121453420 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290837.1 NP_001277766.1 1 - 2 247 117
NM_011354.3 NP_035484.1 1 - 2 247 117
NR_110986.1 1 - 2 247 117
GenBank mRNA AK010749.1 - 1 - 2 79 117
CJ073462.1 - 1 - 2 253 117

View Full Product Details