The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023815.4 | NP_076304.2 | 2 - 2 | 1374 | 129 |
GenBank mRNA | AK028411.1 | - | 2 - 2 | 1375 | 129 |
AK049291.1 | - | 2 - 2 | 1359 | 129 | |
BC017155.1 | - | 2 - 2 | 1392 | 129 |