The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011664.3 | NP_035794.1 | 2 - 2 | 311 | 159 |
GenBank mRNA | AK003190.1 | - | 2 - 2 | 312 | 159 |
AK007273.1 | - | 2 - 2 | 343 | 159 | |
AK010413.1 | - | 2 - 2 | 313 | 159 | |
AK010625.1 | - | 2 - 2 | 312 | 159 | |
AK012443.1 | - | 2 - 2 | 313 | 159 | |
AK013027.1 | - | 2 - 2 | 312 | 159 | |
AK132747.1 | - | 2 - 2 | 312 | 159 | |
AK165313.1 | - | 2 - 2 | 311 | 159 | |
BC019850.1 | - | 2 - 2 | 736 | 159 | |
BC066197.1 | - | 2 - 2 | 341 | 159 | |
BC100341.1 | - | 2 - 2 | 266 | 159 | |
X51703.1 | - | 2 - 2 | 317 | 159 |